The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020407.4 | NP_065140.3 | 1 - 2 | 246 | 71 |
XM_017001857.1 | XP_016857346.1 | 1 - 2 | 232 | 71 | |
XM_017001858.1 | XP_016857347.1 | 1 - 2 | 232 | 71 | |
GenBank mRNA | AF193807.1 | - | 1 - 2 | 223 | 71 |