The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006080.2 | NP_006071.1 | 15 - 16 | 2027 | 120 |
XM_005250110.3 | XP_005250167.1 | 18 - 19 | 2222 | 120 | |
XM_005250111.4 | XP_005250168.1 | 17 - 18 | 2062 | 120 | |
XM_006715839.3 | XP_006715902.1 | 16 - 17 | 1965 | 120 | |
XM_011515734.2 | XP_011514036.1 | 18 - 19 | 2047 | 120 | |
XM_017011673.1 | XP_016867162.1 | 16 - 17 | 1882 | 120 | |
GenBank mRNA | AK289954.1 | - | 15 - 16 | 2120 | 120 |
BC111416.1 | - | 15 - 16 | 1726 | 120 | |
L26081.1 | - | 15 - 16 | 1727 | 120 |