The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005193.1 | NP_005184.1 | 2 - 3 | 642 | 73 |
GenBank mRNA | BC128233.1 | - | 2 - 3 | 642 | 73 |