The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297644.1 | NP_001284573.1 | 6 - 7 | 816 | 85 |
NM_001297645.1 | NP_001284574.1 | 6 - 7 | 816 | 85 | |
NM_001318000.1 | NP_001304929.1 | 6 - 7 | 816 | 85 | |
NM_005869.3 | NP_005860.2 | 6 - 7 | 816 | 85 | |
XM_011543095.2 | XP_011541397.2 | 6 - 7 | 785 | 85 | |
XM_011543096.1 | XP_011541398.1 | 6 - 7 | 785 | 85 | |
XM_017008943.1 | XP_016864432.1 | 6 - 7 | 785 | 85 | |
GenBank mRNA | AF039692.1 | - | 6 - 7 | 768 | 85 |
AF039693.1 | - | 2 - 3 | 207 | 85 | |
AK225105.1 | - | 6 - 7 | 783 | 85 | |
AK225161.1 | - | 6 - 7 | 788 | 85 | |
AK308223.1 | - | 6 - 7 | 785 | 85 | |
AY358569.1 | - | 6 - 7 | 777 | 85 | |
BC012117.1 | - | 6 - 7 | 769 | 85 | |
BX647971.1 | - | 6 - 7 | 817 | 85 |