The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142462.2 | NP_001135934.1 | 3 - 4 | 1292 | 136 |
NM_001286841.1 | NP_001273770.1 | 4 - 5 | 1541 | 136 | |
XM_011516825.2 | XP_011515127.1 | 4 - 5 | 1307 | 136 | |
XM_011516826.2 | XP_011515128.1 | 3 - 4 | 933 | 136 | |
XM_011516827.2 | XP_011515129.1 | 3 - 4 | 1040 | 136 | |
XM_017013018.1 | XP_016868507.1 | 3 - 4 | 912 | 136 | |
GenBank mRNA | AK074518.1 | - | 3 - 4 | 1253 | 136 |
AK304654.1 | - | 4 - 5 | 1285 | 136 | |
AY038072.1 | - | 3 - 4 | 1072 | 136 |