The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021223.2 | NP_067046.1 | 2 - 3 | 209 | 84 |
XM_005249817.4 | XP_005249874.1 | 3 - 4 | 352 | 84 | |
XM_011515463.2 | XP_011513765.1 | 3 - 4 | 353 | 84 | |
XM_011515464.2 | XP_011513766.1 | 3 - 4 | 283 | 84 | |
XM_011515465.2 | XP_011513767.1 | 3 - 4 | 283 | 84 | |
XM_017012478.1 | XP_016867967.1 | 3 - 4 | 282 | 84 | |
XM_017012479.1 | XP_016867968.1 | 3 - 4 | 290 | 84 | |
GenBank mRNA | AK311869.1 | - | 3 - 4 | 224 | 84 |
BC027915.1 | - | 2 - 3 | 205 | 84 | |
M94547.1 | - | 2 - 3 | 209 | 84 | |
W17098.1 | - | 3 - 4 | 210 | 84 |