The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166292.1 | NP_001159764.1 | 9 - 10 | 1228 | 57 |
NM_003738.4 | NP_003729.3 | 9 - 10 | 1228 | 57 | |
GenBank mRNA | AF087651.1 | - | 9 - 10 | 1216 | 57 |
AF091501.1 | - | 9 - 10 | 1216 | 57 | |
AF119569.1 | - | 9 - 10 | 1228 | 57 | |
AY358555.1 | - | 9 - 10 | 1216 | 57 | |
AY359016.1 | - | 9 - 10 | 1220 | 57 | |
BC143712.1 | - | 8 - 9 | 1088 | 57 |