The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002444.2 | NP_002435.1 | 2 - 3 | 288 | 63 |
XM_005262269.2 | XP_005262326.1 | 2 - 3 | 309 | 63 | |
XM_011530959.1 | XP_011529261.1 | 2 - 3 | 204 | 63 | |
XM_017029545.1 | XP_016885034.1 | 4 - 5 | 432 | 63 | |
XM_017029546.1 | XP_016885035.1 | 3 - 4 | 328 | 63 | |
GenBank mRNA | AK308486.1 | - | 2 - 3 | 251 | 63 |
AK309354.1 | - | 1 - 2 | 53 | 63 | |
BC001112.2 | - | 2 - 3 | 260 | 63 | |
BC011827.2 | - | 2 - 3 | 260 | 63 | |
BC017293.2 | - | 2 - 3 | 241 | 63 | |
CN386296.1 | - | 2 - 3 | 288 | 63 | |
EU794636.1 | - | 2 - 3 | 241 | 63 | |
M69066.1 | - | 2 - 3 | 190 | 63 |