The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321386.1 | NP_001308315.1 | 2 - 3 | 231 | 70 |
NM_001321387.1 | NP_001308316.1 | 2 - 3 | 533 | 70 | |
NM_002825.5 | NP_002816.1 | 2 - 3 | 468 | 70 | |
XM_017012473.1 | XP_016867962.1 | 2 - 3 | 351 | 70 | |
GenBank mRNA | AK290488.1 | - | 2 - 3 | 297 | 70 |
AK313424.1 | - | 2 - 3 | 371 | 70 | |
BC005916.1 | - | 2 - 3 | 456 | 70 | |
BT019692.1 | - | 1 - 2 | 105 | 70 | |
BX420480.2 | - | 2 - 3 | 532 | 70 | |
BX441769.2 | - | 2 - 3 | 236 | 70 | |
CN413530.1 | - | 2 - 3 | 217 | 70 | |
CR450338.1 | - | 1 - 2 | 105 | 70 | |
D90226.1 | - | 2 - 3 | 229 | 70 | |
M57399.1 | - | 2 - 3 | 500 | 70 | |
X52946.1 | - | 2 - 3 | 356 | 70 |