Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GATA1
Entrez Gene ID
2623
Gene Name
GATA binding protein 1
Gene Aliases
ERYF1, GATA-1, GF-1, GF1, NF-E1, NFE1, XLANP, XLTDA, XLTT
Chromosome Location
Chr.X: 48786540 - 48794311 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002049.3 NP_002040.1 3 - 4 691 71
XM_011543897.2 XP_011542199.1 3 - 4 725 71
XM_011543898.2 XP_011542200.1 2 - 3 485 71
GenBank mRNA BC009797.1 - 3 - 4 694 71
M30601.1 - 3 - 4 658 71
X17254.1 - 3 - 4 712 71

View Full Product Details