Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNG
Entrez Gene ID
55787
Gene Name
taxilin gamma
Gene Aliases
CXorf15, ELRG, FIAT, LSR5, TXLNGX
Chromosome Location
Chr.X: 16786432 - 16844519 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018360.2 NP_060830.2 1 - 2 165 96
GenBank mRNA AK002071.1 - 1 - 2 160 96
BC101572.1 - 1 - 2 132 96
BC101576.1 - 1 - 2 132 96
DB098819.1 - 1 - 2 165 96

View Full Product Details