The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000920.3 | NP_000911.2 | 7 - 8 | 845 | 60 |
NM_001040716.1 | NP_001035806.1 | 8 - 9 | 1033 | 60 | |
NM_022172.2 | NP_071504.2 | 6 - 7 | 800 | 60 | |
XM_005274031.4 | XP_005274088.1 | 6 - 7 | 892 | 60 | |
XM_005274032.4 | XP_005274089.1 | 6 - 7 | 774 | 60 | |
XM_006718578.3 | XP_006718641.1 | 6 - 7 | 845 | 60 | |
XM_011545086.2 | XP_011543388.1 | 6 - 7 | 1161 | 60 | |
XM_017017868.1 | XP_016873357.1 | 9 - 10 | 1332 | 60 | |
XM_017017869.1 | XP_016873358.1 | 9 - 10 | 1203 | 60 | |
XM_017017870.1 | XP_016873359.1 | 8 - 9 | 1144 | 60 | |
XM_017017871.1 | XP_016873360.1 | 8 - 9 | 1015 | 60 | |
XM_017017872.1 | XP_016873361.1 | 7 - 8 | 1397 | 60 | |
GenBank mRNA | AK297705.1 | - | 7 - 8 | 845 | 60 |
BC011617.2 | - | 7 - 8 | 834 | 60 | |
S72370.1 | - | 6 - 7 | 833 | 60 | |
U04641.1 | - | 6 - 7 | 789 | 60 | |
U30891.1 | - | 7 - 8 | 803 | 60 |