The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129896.2 | NP_001123368.1 | 7 - 8 | 952 | 84 |
NM_001174068.1 | NP_001167539.1 | 6 - 7 | 831 | 84 | |
NM_080737.2 | NP_542775.2 | 8 - 9 | 1000 | 84 | |
XM_005262229.2 | XP_005262286.1 | 6 - 7 | 832 | 84 | |
XM_011531068.2 | XP_011529370.1 | 8 - 9 | 1029 | 84 | |
XM_017029966.1 | XP_016885455.1 | 8 - 9 | 1029 | 84 | |
XM_017029967.1 | XP_016885456.1 | 7 - 8 | 921 | 84 | |
XM_017029968.1 | XP_016885457.1 | 6 - 7 | 832 | 84 | |
XM_017029969.1 | XP_016885458.1 | 6 - 7 | 806 | 84 | |
XM_017029970.1 | XP_016885459.1 | 6 - 7 | 754 | 84 | |
XM_017029971.1 | XP_016885460.1 | 7 - 8 | 932 | 84 | |
GenBank mRNA | AF302493.1 | - | 5 - 6 | 717 | 84 |
AK292588.1 | - | 7 - 8 | 921 | 84 | |
AK313086.1 | - | 6 - 7 | 806 | 84 | |
AL832596.1 | - | 6 - 7 | 831 | 84 | |
BC014913.1 | - | 8 - 9 | 976 | 84 | |
BX537410.1 | - | 7 - 8 | 919 | 84 |