Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCRS1
Entrez Gene ID
10445
Gene Name
microspherule protein 1
Gene Aliases
ICP22BP, INO80Q, MCRS2, MSP58, P78
Chromosome Location
Chr.12: 49558294 - 49572157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012300.1 NP_001012300.1 6 - 7 709 66
NM_001278341.1 NP_001265270.1 5 - 6 619 66
NM_006337.4 NP_006328.2 7 - 8 878 66
XM_005268572.3 XP_005268629.1 8 - 9 978 66
XM_011537760.2 XP_011536062.1 6 - 7 4770 66
XM_017018689.1 XP_016874178.1 7 - 8 1214 66
XM_017018690.1 XP_016874179.1 8 - 9 1299 66
GenBank mRNA AF015308.1 - 6 - 7 786 66
AF068007.1 - 6 - 7 1418 66
AY336730.1 - 6 - 7 709 66
BC011794.2 - 7 - 8 861 66
BX538079.1 - 5 - 6 606 66

View Full Product Details