Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFS8
Entrez Gene ID
4728
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
Gene Aliases
CI-23k, CI23KD, TYKY
Chromosome Location
Chr.11: 68030617 - 68036647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002496.3 NP_002487.1 3 - 4 223 127
XM_005274013.1 XP_005274070.1 3 - 4 320 127
XM_005274014.2 XP_005274071.1 3 - 4 347 127
XM_011545053.2 XP_011543355.1 3 - 4 258 127
GenBank mRNA AK223114.1 - 3 - 4 213 127
AK302449.1 - 3 - 4 223 127
AK314546.1 - 3 - 4 148 127
BC119754.2 - 3 - 4 152 127
DB498707.1 - 3 - 4 209 127
U65579.1 - 3 - 4 146 127

View Full Product Details