The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271618.1 | NP_001258547.1 | 6 - 7 | 1014 | 57 |
NM_017607.3 | NP_060077.1 | 6 - 7 | 1014 | 57 | |
XM_005259013.3 | XP_005259070.1 | 6 - 7 | 1005 | 57 | |
XM_011527045.2 | XP_011525347.1 | 6 - 7 | 1006 | 57 | |
GenBank mRNA | AB209452.1 | - | 6 - 7 | 827 | 57 |
AF312028.1 | - | 6 - 7 | 975 | 57 | |
AK027086.1 | - | 6 - 7 | 652 | 57 | |
AK297187.1 | - | 3 - 4 | 300 | 57 | |
AK297426.1 | - | 6 - 7 | 863 | 57 | |
AL137618.1 | - | 1 - 2 | 48 | 57 | |
BC010628.1 | - | 5 - 6 | 792 | 57 |