The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017607.3 | NP_060077.1 | 9 - 10 | 1285 | 74 |
XM_005259013.3 | XP_005259070.1 | 9 - 10 | 1276 | 74 | |
GenBank mRNA | AF312028.1 | - | 9 - 10 | 1246 | 74 |
AK297426.1 | - | 9 - 10 | 1134 | 74 | |
AL137618.1 | - | 4 - 5 | 319 | 74 | |
BC010628.1 | - | 8 - 9 | 1063 | 74 |