The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015974.2 | NP_057058.2 | 5 - 6 | 712 | 82 |
XM_005266415.3 | XP_005266472.1 | 4 - 5 | 539 | 82 | |
XM_017020624.1 | XP_016876113.1 | 6 - 7 | 769 | 82 | |
GenBank mRNA | AF087898.1 | - | 6 - 7 | 739 | 82 |
AF160216.1 | - | 5 - 6 | 640 | 82 | |
AK024041.1 | - | 5 - 6 | 696 | 82 | |
BC008562.2 | - | 4 - 5 | 594 | 82 | |
BC071810.1 | - | 6 - 7 | 943 | 82 | |
BC119660.1 | - | 5 - 6 | 615 | 82 | |
BC119661.1 | - | 5 - 6 | 615 | 82 | |
EU794604.1 | - | 6 - 7 | 739 | 82 |