The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077446.3 | NP_001070914.1 | 1 - 2 | 395 | 58 |
NM_001282332.1 | NP_001269261.1 | 2 - 3 | 555 | 58 | |
NM_001282333.1 | NP_001269262.1 | 2 - 3 | 352 | 58 | |
NM_024075.4 | NP_076980.2 | 2 - 3 | 431 | 58 | |
XM_005278290.4 | XP_005278347.1 | 2 - 3 | 347 | 58 | |
XM_011527294.2 | XP_011525596.1 | 2 - 3 | 326 | 58 | |
XM_011527295.1 | XP_011525597.1 | 2 - 3 | 328 | 58 | |
GenBank mRNA | AK025929.1 | - | 2 - 3 | 350 | 58 |
AK225385.1 | - | 2 - 3 | 350 | 58 | |
AK226013.1 | - | 2 - 3 | 353 | 58 | |
AK300054.1 | - | 2 - 3 | 333 | 58 | |
BC004530.2 | - | 2 - 3 | 555 | 58 | |
BC020805.1 | - | 1 - 2 | 335 | 58 | |
DR763016.1 | - | 2 - 3 | 327 | 58 |