The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127197.1 | NP_001120669.1 | 7 - 8 | 1113 | 82 |
NM_001421.3 | NP_001412.1 | 7 - 8 | 1195 | 82 | |
XM_005262389.3 | XP_005262446.1 | 7 - 8 | 1531 | 82 | |
XM_011531307.2 | XP_011529609.1 | 6 - 7 | 941 | 82 | |
XM_011531308.2 | XP_011529610.1 | 5 - 6 | 723 | 82 | |
GenBank mRNA | AF000670.1 | - | 6 - 7 | 912 | 82 |
AK314972.1 | - | 7 - 8 | 1112 | 82 | |
BC017194.2 | - | 7 - 8 | 1078 | 82 | |
U32645.1 | - | 7 - 8 | 1192 | 82 |