The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000695.3 | NP_000686.2 | 9 - 10 | 1411 | 67 |
NM_001031615.1 | NP_001026786.1 | 9 - 10 | 1255 | 67 | |
GenBank mRNA | AK092464.1 | - | 9 - 10 | 1468 | 67 |
AK299956.1 | - | 4 - 5 | 736 | 67 | |
BC007685.2 | - | 9 - 10 | 1255 | 67 | |
BT006810.1 | - | 7 - 8 | 972 | 67 |