The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168278.1 | NP_001161750.1 | 7 - 8 | 1276 | 72 |
NM_001168280.1 | NP_001161752.1 | 6 - 7 | 1223 | 72 | |
NM_015472.4 | NP_056287.1 | 6 - 7 | 1359 | 72 | |
XM_017006121.1 | XP_016861610.1 | 9 - 10 | 1469 | 72 | |
XM_017006122.1 | XP_016861611.1 | 7 - 8 | 1268 | 72 | |
GenBank mRNA | AJ299431.1 | - | 6 - 7 | 1194 | 72 |
AK022036.1 | - | 7 - 8 | 1276 | 72 | |
AL050107.1 | - | 4 - 5 | 583 | 72 | |
AL833852.1 | - | 5 - 6 | 1013 | 72 | |
BC014052.2 | - | 6 - 7 | 1253 | 72 |