Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
55345
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C4orf21
Chromosome Location
Chr.4: 112539333 - 112636995 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018392.4 NP_060862.3 19 - 20 5175 71
XM_005263115.3 XP_005263172.1 19 - 20 5230 71
XM_011532091.2 XP_011530393.1 18 - 19 5124 71
XM_011532092.2 XP_011530394.1 18 - 19 5055 71
XM_011532093.2 XP_011530395.1 18 - 19 5001 71
XM_011532094.2 XP_011530396.1 17 - 18 4917 71
XM_011532097.2 XP_011530399.1 12 - 13 2277 71
XM_011532098.2 XP_011530400.1 10 - 11 1951 71
XM_011532099.2 XP_011530401.1 9 - 10 1910 71
XM_017008369.1 XP_016863858.1 18 - 19 5170 71
XM_017008370.1 XP_016863859.1 17 - 18 4950 71
XM_017008371.1 XP_016863860.1 16 - 17 4830 71
XM_017008372.1 XP_016863861.1 16 - 17 4209 71
XM_017008373.1 XP_016863862.1 15 - 16 4146 71
GenBank mRNA AK090556.1 - 1 - 2 810 71
AK126054.1 - 8 - 9 1559 71
AL137700.1 - 5 - 6 750 71
BX647594.1 - 8 - 9 1658 71

View Full Product Details