The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152540.3 | NP_689753.2 | 3 - 4 | 1267 | 63 |
XM_011534375.2 | XP_011532677.1 | 3 - 4 | 1303 | 63 | |
XM_011534376.2 | XP_011532678.1 | 3 - 4 | 1303 | 63 | |
XM_011534378.2 | XP_011532680.1 | 3 - 4 | 1303 | 63 | |
XM_017007786.1 | XP_016863275.1 | 3 - 4 | 1303 | 63 | |
XM_017007787.1 | XP_016863276.1 | 3 - 4 | 1303 | 63 | |
GenBank mRNA | AB074276.1 | - | 3 - 4 | 1240 | 63 |
AK096833.1 | - | 3 - 4 | 1253 | 63 | |
AY299407.1 | - | 3 - 4 | 1267 | 63 | |
BC012458.1 | - | 3 - 4 | 1033 | 63 | |
BC021137.1 | - | 3 - 4 | 1224 | 63 | |
BC032453.1 | - | 3 - 4 | 1265 | 63 |