The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152540.3 | NP_689753.2 | 7 - 8 | 1976 | 101 |
GenBank mRNA | AB074276.1 | - | 7 - 8 | 1949 | 101 |
AK024713.1 | - | 1 - 2 | 113 | 101 | |
AY299407.1 | - | 7 - 8 | 1976 | 101 | |
BC012458.1 | - | 7 - 8 | 1742 | 101 | |
BC021137.1 | - | 7 - 8 | 1933 | 101 | |
BC032453.1 | - | 7 - 8 | 1974 | 101 |