The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163508.1 | NP_001156980.1 | 3 - 4 | 413 | 104 |
NM_015604.3 | NP_056419.2 | 3 - 4 | 413 | 104 | |
NM_181340.2 | NP_851937.1 | 2 - 3 | 313 | 104 | |
NM_181341.2 | NP_851938.2 | 3 - 4 | 413 | 104 | |
XM_017021201.1 | XP_016876690.1 | 2 - 3 | 222 | 104 | |
XM_017021202.1 | XP_016876691.1 | 2 - 3 | 222 | 104 | |
XM_017021203.1 | XP_016876692.1 | 3 - 4 | 408 | 104 | |
XM_017021204.1 | XP_016876693.1 | 3 - 4 | 378 | 104 | |
XM_017021205.1 | XP_016876694.1 | 4 - 5 | 547 | 104 | |
XM_017021206.1 | XP_016876695.1 | 3 - 4 | 383 | 104 | |
XM_017021207.1 | XP_016876696.1 | 3 - 4 | 408 | 104 | |
XM_017021208.1 | XP_016876697.1 | 3 - 4 | 408 | 104 | |
XM_017021209.1 | XP_016876698.1 | 3 - 4 | 408 | 104 | |
XM_017021210.1 | XP_016876699.1 | 3 - 4 | 408 | 104 | |
XM_017021215.1 | XP_016876704.1 | 3 - 4 | 382 | 104 | |
GenBank mRNA | AK300786.1 | - | 3 - 4 | 265 | 104 |
AK315499.1 | - | 3 - 4 | 413 | 104 | |
AL080157.1 | - | 2 - 3 | 184 | 104 | |
BC018979.2 | - | 3 - 4 | 225 | 104 | |
BC035901.1 | - | 2 - 3 | 167 | 104 | |
BX161489.1 | - | 3 - 4 | 368 | 104 | |
BX247962.1 | - | 3 - 4 | 240 | 104 | |
DC314513.1 | - | 3 - 4 | 413 | 104 |