Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYZ
Entrez Gene ID
1429
Gene Name
crystallin zeta
Gene Aliases
-
Chromosome Location
Chr.1: 74705482 - 74733408 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130042.1 NP_001123514.1 6 - 7 982 104
NM_001130043.1 NP_001123515.1 5 - 6 943 104
NM_001134759.1 NP_001128231.1 3 - 4 655 104
NM_001889.3 NP_001880.2 5 - 6 943 104
XM_005270491.4 XP_005270548.1 5 - 6 456 104
XM_011540747.1 XP_011539049.1 5 - 6 562 104
XM_017000367.1 XP_016855856.1 6 - 7 617 104
XM_017000368.1 XP_016855857.1 4 - 5 466 104
GenBank mRNA AB209714.1 - 6 - 7 614 104
AK128794.1 - 5 - 6 548 104
AK223150.1 - 5 - 6 575 104
AK223201.1 - 5 - 6 575 104
AK314813.1 - 5 - 6 583 104
BC039578.1 - 5 - 6 565 104
BC070058.1 - 6 - 7 624 104
BX647883.1 - 3 - 4 266 104
L13278.1 - 4 - 5 487 104

View Full Product Details