The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008744.1 | NP_001008744.1 | 12 - 13 | 1512 | 83 |
NM_018319.3 | NP_060789.2 | 13 - 14 | 1735 | 83 | |
XM_005267847.2 | XP_005267904.1 | 13 - 14 | 2409 | 83 | |
XM_005267848.2 | XP_005267905.1 | 12 - 13 | 1753 | 83 | |
XM_006720197.3 | XP_006720260.1 | 13 - 14 | 1976 | 83 | |
XM_006720198.3 | XP_006720261.1 | 12 - 13 | 1631 | 83 | |
XM_006720199.1 | XP_006720262.1 | 12 - 13 | 1117 | 83 | |
XM_011536942.2 | XP_011535244.1 | 12 - 13 | 2152 | 83 | |
XM_011536943.2 | XP_011535245.1 | 12 - 13 | 1545 | 83 | |
XM_011536944.1 | XP_011535246.1 | 13 - 14 | 1736 | 83 | |
XM_017021439.1 | XP_016876928.1 | 13 - 14 | 1643 | 83 | |
XM_017021440.1 | XP_016876929.1 | 12 - 13 | 1535 | 83 | |
XM_017021441.1 | XP_016876930.1 | 12 - 13 | 1724 | 83 | |
XM_017021442.1 | XP_016876931.1 | 10 - 11 | 1361 | 83 | |
XM_017021443.1 | XP_016876932.1 | 10 - 11 | 860 | 83 | |
XM_017021444.1 | XP_016876933.1 | 10 - 11 | 861 | 83 | |
XM_017021445.1 | XP_016876934.1 | 10 - 11 | 1361 | 83 | |
XM_017021446.1 | XP_016876935.1 | 11 - 12 | 1167 | 83 | |
GenBank mRNA | AF182003.1 | - | 3 - 4 | 181 | 83 |
AK001952.1 | - | 12 - 13 | 1476 | 83 | |
AK023514.1 | - | 2 - 3 | 98 | 83 | |
AK093235.1 | - | 14 - 15 | 2543 | 83 | |
AK315549.1 | - | 12 - 13 | 1460 | 83 | |
AL832288.1 | - | 4 - 5 | 454 | 83 | |
BC006083.1 | - | 15 - 16 | 1899 | 83 | |
BC015474.1 | - | 13 - 14 | 1686 | 83 | |
BC107877.1 | - | 13 - 14 | 1739 | 83 | |
BX161451.1 | - | 12 - 13 | 1647 | 83 |