Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDP1
Entrez Gene ID
55775
Gene Name
tyrosyl-DNA phosphodiesterase 1
Gene Aliases
-
Chromosome Location
Chr.14: 89954935 - 90044764 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008744.1 NP_001008744.1 9 - 10 1213 151
NM_018319.3 NP_060789.2 10 - 11 1436 151
XM_005267847.2 XP_005267904.1 10 - 11 2110 151
XM_005267848.2 XP_005267905.1 9 - 10 1454 151
XM_006720197.3 XP_006720260.1 10 - 11 1677 151
XM_006720198.3 XP_006720261.1 9 - 10 1332 151
XM_006720199.1 XP_006720262.1 9 - 10 818 151
XM_011536942.2 XP_011535244.1 9 - 10 1853 151
XM_011536943.2 XP_011535245.1 9 - 10 1246 151
XM_011536944.1 XP_011535246.1 10 - 11 1437 151
XM_017021439.1 XP_016876928.1 10 - 11 1344 151
XM_017021440.1 XP_016876929.1 9 - 10 1236 151
XM_017021441.1 XP_016876930.1 9 - 10 1425 151
XM_017021442.1 XP_016876931.1 7 - 8 1062 151
XM_017021443.1 XP_016876932.1 7 - 8 561 151
XM_017021444.1 XP_016876933.1 7 - 8 562 151
XM_017021445.1 XP_016876934.1 7 - 8 1062 151
XM_017021446.1 XP_016876935.1 8 - 9 868 151
GenBank mRNA AF182002.1 - 8 - 9 1122 151
AK093235.1 - 11 - 12 2244 151
AK315549.1 - 9 - 10 1161 151
BC015474.1 - 10 - 11 1387 151
BC107877.1 - 10 - 11 1440 151
BX161451.1 - 9 - 10 1348 151

View Full Product Details