The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169.2 | NP_001160.2 | 5 - 6 | 819 | 60 |
XM_011545822.2 | XP_011544124.1 | 5 - 6 | 862 | 60 | |
XM_011545823.2 | XP_011544125.1 | 3 - 4 | 527 | 60 | |
GenBank mRNA | AB013456.1 | - | 5 - 6 | 821 | 60 |
AF067797.1 | - | 5 - 6 | 822 | 60 | |
AK222964.1 | - | 5 - 6 | 784 | 60 | |
AK222975.1 | - | 5 - 6 | 816 | 60 | |
BC040630.1 | - | 5 - 6 | 787 | 60 | |
BG221545.1 | - | 1 - 2 | 139 | 60 |