The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243766.1 | NP_001230695.1 | 5 - 6 | 1070 | 78 |
NM_001290129.1 | NP_001277058.1 | 4 - 5 | 491 | 78 | |
NM_001290130.1 | NP_001277059.1 | 4 - 5 | 457 | 78 | |
NM_017739.3 | NP_060209.3 | 5 - 6 | 583 | 78 | |
XM_005271010.1 | XP_005271067.1 | 5 - 6 | 578 | 78 | |
XM_006710755.1 | XP_006710818.1 | 5 - 6 | 543 | 78 | |
XM_006710756.1 | XP_006710819.1 | 5 - 6 | 578 | 78 | |
XM_011541760.2 | XP_011540062.1 | 4 - 5 | 435 | 78 | |
XM_017001690.1 | XP_016857179.1 | 5 - 6 | 543 | 78 | |
GenBank mRNA | AB057356.1 | - | 5 - 6 | 513 | 78 |
AF250859.2 | - | 2 - 3 | 172 | 78 | |
AK000284.1 | - | 5 - 6 | 560 | 78 | |
AK026430.1 | - | - | 2151 | 78 | |
AK056186.1 | - | 5 - 6 | 1070 | 78 | |
AK225033.1 | - | 5 - 6 | 561 | 78 | |
AK301923.1 | - | 4 - 5 | 457 | 78 | |
AK302379.1 | - | 4 - 5 | 467 | 78 | |
AK316399.1 | - | 4 - 5 | 464 | 78 | |
AY358592.1 | - | 5 - 6 | 548 | 78 | |
BC001471.2 | - | 5 - 6 | 577 | 78 | |
CR749700.1 | - | 5 - 6 | 1056 | 78 |