The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130080.2 | NP_001123552.1 | 3 - 4 | 248 | 79 |
NM_001288952.1 | NP_001275881.1 | 4 - 5 | 301 | 79 | |
NM_001288956.1 | NP_001275885.1 | 3 - 4 | 214 | 79 | |
NM_001288959.1 | NP_001275888.1 | 2 - 3 | 99 | 79 | |
GenBank mRNA | AJ294851.2 | - | 3 - 4 | 209 | 79 |
AK289535.1 | - | 3 - 4 | 246 | 79 | |
AK312089.1 | - | 3 - 4 | 245 | 79 | |
AY126457.1 | - | 2 - 3 | 93 | 79 | |
BF971643.1 | - | 4 - 5 | 280 | 79 | |
BG483267.1 | - | 3 - 4 | 212 | 79 | |
BU601451.1 | - | 3 - 4 | 248 | 79 | |
BU688913.1 | - | 2 - 3 | 79 | ||
BU928223.1 | - | 2 - 3 | 100 | 79 | |
BX647330.1 | - | 3 - 4 | 245 | 79 | |
X67325.1 | - | 2 - 3 | 179 | 79 |