The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015557.2 | NP_056372.1 | 5 - 6 | 859 | 63 |
GenBank mRNA | AF425231.1 | - | 5 - 6 | 848 | 63 |
AK127046.1 | - | 5 - 6 | 748 | 63 |