The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077238.1 | NP_001070706.1 | 7 - 8 | 918 | 70 |
NM_152988.2 | NP_694533.1 | 7 - 8 | 918 | 70 | |
XM_011528138.2 | XP_011526440.1 | 7 - 8 | 974 | 70 | |
XM_011528139.1 | XP_011526441.1 | 6 - 7 | 776 | 70 | |
XM_011528140.1 | XP_011526442.1 | 3 - 4 | 329 | 70 | |
XM_017027017.1 | XP_016882506.1 | 8 - 9 | 1112 | 70 | |
XM_017027018.1 | XP_016882507.1 | 6 - 7 | 773 | 70 | |
XM_017027019.1 | XP_016882508.1 | 8 - 9 | 1105 | 70 | |
GenBank mRNA | AB040965.1 | - | 7 - 8 | 867 | 70 |
AJ345027.1 | - | 7 - 8 | 871 | 70 | |
AJ420897.1 | - | 7 - 8 | 867 | 70 | |
AK226130.1 | - | 4 - 5 | 458 | 70 | |
AK308973.1 | - | 8 - 9 | 987 | 70 | |
AK309122.1 | - | 7 - 8 | 992 | 70 | |
AY169315.1 | - | 7 - 8 | 866 | 70 | |
BC001788.1 | - | 7 - 8 | 852 | 70 | |
BC028391.2 | - | 7 - 8 | 855 | 70 | |
BC093046.1 | - | 7 - 8 | 918 | 70 |