The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002096.2 | NP_002087.2 | 6 - 7 | 966 | 70 |
GenBank mRNA | AK302178.1 | - | 3 - 4 | 429 | 70 |
AK315240.1 | - | 6 - 7 | 850 | 70 | |
BC000120.1 | - | 6 - 7 | 828 | 70 | |
BC013007.2 | - | 6 - 7 | 828 | 70 | |
BT007097.1 | - | 5 - 6 | 681 | 70 | |
CR456798.1 | - | 5 - 6 | 681 | 70 | |
X64037.1 | - | 6 - 7 | 859 | 70 |