The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048201.2 | NP_001041666.1 | 4 - 5 | 804 | 116 |
NM_001290050.1 | NP_001276979.1 | 4 - 5 | 1144 | 116 | |
NM_001290051.1 | NP_001276980.1 | 4 - 5 | 732 | 116 | |
NM_001290052.1 | NP_001276981.1 | 4 - 5 | 706 | 116 | |
NM_013282.4 | NP_037414.3 | 3 - 4 | 1087 | 116 | |
XM_011527942.2 | XP_011526244.1 | 4 - 5 | 733 | 116 | |
GenBank mRNA | AB177623.1 | - | 4 - 5 | 1144 | 116 |
AB177624.1 | - | 3 - 4 | 1087 | 116 | |
AF129507.1 | - | 3 - 4 | 568 | 116 | |
AF274048.1 | - | 3 - 4 | 652 | 116 | |
AK074377.1 | - | 4 - 5 | 733 | 116 | |
AK289389.1 | - | 4 - 5 | 645 | 116 | |
AK314579.1 | - | 4 - 5 | 733 | 116 | |
BC113875.1 | - | 3 - 4 | 652 | 116 | |
EF560733.1 | - | 4 - 5 | 699 | 116 |