The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006769.3 | NP_006760.1 | 2 - 3 | 1018 | 65 |
XM_005271291.3 | XP_005271348.1 | 2 - 3 | 3188 | 65 | |
GenBank mRNA | AK292578.1 | - | 2 - 3 | 620 | 65 |
BC003600.1 | - | 2 - 3 | 381 | 65 | |
BC017673.1 | - | 2 - 3 | 506 | 65 | |
BC065818.1 | - | 2 - 3 | 312 | 65 | |
U24576.1 | - | 2 - 3 | 1018 | 65 |