Product Details

Assay Reference Genome
Location

Chr.14:75006590 on build GRCh38
Cytoband
14q24.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8892
Gene Symbol
EIF2B2
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
Location
Chr.14:75002909-75009591 on build GRCh38
Assay Gene Location
Overlaps Intron 5 - Exon 6
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
EIF2B2 NM_014239.3 NP_055054.1
AF035280.1 AAB88176.1
BC000494.2 AAH00494.1
BC003165.1 AAH03165.1
BC011750.2 AAH11750.1
BI908906.1
BX161473.1 CAD61929.1
L40395.1 AAC42002.1

View Full Product Details