The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321783.1 | NP_001308712.1 | 20 - 21 | 7872 | 156 |
NM_001321784.1 | NP_001308713.1 | 20 - 21 | 7918 | 156 | |
NM_001321785.1 | NP_001308714.1 | 19 - 20 | 7629 | 156 | |
NM_017782.4 | NP_060252.4 | 19 - 20 | 7799 | 156 | |
XM_005252474.3 | XP_005252531.1 | 22 - 23 | 14610 | 156 | |
XM_005252475.3 | XP_005252532.1 | 23 - 24 | 8271 | 156 | |
XM_005252476.4 | XP_005252533.1 | 21 - 22 | 8054 | 156 | |
XM_005252480.4 | XP_005252537.1 | 19 - 20 | 7859 | 156 | |
XM_005252481.4 | XP_005252538.1 | 17 - 18 | 7344 | 156 | |
XM_005252482.2 | XP_005252539.1 | 21 - 22 | 7704 | 156 | |
XM_011519515.2 | XP_011517817.1 | 23 - 24 | 14748 | 156 | |
XM_011519517.2 | XP_011517819.1 | 23 - 24 | 8668 | 156 | |
XM_011519518.2 | XP_011517820.1 | 22 - 23 | 7981 | 156 | |
XM_011519519.2 | XP_011517821.1 | 22 - 23 | 14505 | 156 | |
XM_011519520.2 | XP_011517822.1 | 20 - 21 | 14445 | 156 | |
XM_011519521.2 | XP_011517823.1 | 22 - 23 | 8192 | 156 | |
XM_011519522.2 | XP_011517824.1 | 22 - 23 | 8576 | 156 | |
XM_011519525.2 | XP_011517827.1 | 20 - 21 | 7612 | 156 | |
XM_017016362.1 | XP_016871851.1 | 21 - 22 | 8475 | 156 | |
XM_017016363.1 | XP_016871852.1 | 22 - 23 | 8133 | 156 | |
XM_017016364.1 | XP_016871853.1 | 20 - 21 | 8337 | 156 | |
XM_017016365.1 | XP_016871854.1 | 21 - 22 | 7999 | 156 | |
GenBank mRNA | AB095927.1 | - | 6 - 7 | 3987 | 156 |
AK000367.1 | - | 5 - 6 | 2186 | 156 | |
AK022966.1 | - | 5 - 6 | 1705 | 156 | |
AK027014.1 | - | 5 - 6 | 1365 | 156 | |
AK091392.1 | - | 5 - 6 | 2918 | 156 | |
AK128051.1 | - | 7 - 8 | 5074 | 156 | |
AK128055.1 | - | 5 - 6 | 3494 | 156 | |
AK128151.1 | - | 5 - 6 | 3019 | 156 | |
AK225352.1 | - | 5 - 6 | 2186 | 156 | |
AL832762.1 | - | 6 - 7 | 4502 | 156 | |
BC001759.1 | - | 5 - 6 | 2192 | 156 | |
BC110339.1 | - | 5 - 6 | 824 | 156 | |
CR936780.1 | - | 19 - 20 | 7800 | 156 |