The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020988.2 | NP_066268.1 | 5 - 6 | 1488 | 80 |
NM_138736.2 | NP_620073.2 | 5 - 6 | 1488 | 80 | |
XM_011523003.2 | XP_011521305.1 | 5 - 6 | 846 | 80 | |
GenBank mRNA | AF493894.1 | - | 5 - 6 | 591 | 80 |
AF493895.1 | - | 5 - 6 | 591 | 80 | |
AK056008.1 | - | 5 - 6 | 1437 | 80 | |
BC030027.2 | - | 5 - 6 | 524 | 80 | |
CR936770.1 | - | 5 - 6 | 851 | 80 | |
KR081949.1 | - | 5 - 6 | 591 | 80 | |
M19182.1 | - | 3 - 4 | 431 | 80 |