Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNAO1
Entrez Gene ID
2775
Gene Name
G protein subunit alpha o1
Gene Aliases
EIEE17, G-ALPHA-o, GNAO, HLA-DQB1
Chromosome Location
Chr.16: 56191339 - 56357444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138736.2 NP_620073.2 6 - 7 1620 80
GenBank mRNA AF493895.1 - 6 - 7 723 80
CR936770.1 - 6 - 7 983 80
KR081949.1 - 6 - 7 723 80

View Full Product Details