The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030775.2 | NP_110402.2 | 4 - 5 | 767 | 65 |
NM_032642.2 | NP_116031.1 | 4 - 5 | 805 | 65 | |
GenBank mRNA | AB060966.1 | - | 3 - 4 | 627 | 65 |
AB209228.1 | - | 3 - 4 | 1165 | 65 | |
AK290430.1 | - | 4 - 5 | 866 | 65 | |
AY009399.1 | - | 4 - 5 | 767 | 65 | |
BC001749.1 | - | 4 - 5 | 805 | 65 |