The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286521.1 | NP_001273450.1 | 3 - 4 | 443 | 98 |
NM_001286522.1 | NP_001273451.1 | 4 - 5 | 565 | 98 | |
NM_020367.5 | NP_065100.2 | 4 - 5 | 564 | 98 | |
NR_104461.1 | 4 - 5 | 564 | 98 | ||
XM_005253711.4 | XP_005253768.1 | 4 - 5 | 958 | 98 | |
XM_011520970.1 | XP_011519272.1 | 5 - 6 | 680 | 98 | |
XM_011520971.2 | XP_011519273.1 | 5 - 6 | 1080 | 98 | |
XM_011520973.2 | XP_011519275.1 | 4 - 5 | 393 | 98 | |
XM_017019670.1 | XP_016875159.1 | 5 - 6 | 602 | 98 | |
XM_017019671.1 | XP_016875160.1 | 4 - 5 | 959 | 98 | |
GenBank mRNA | AF263540.1 | - | 3 - 4 | 379 | 98 |
AK299372.1 | - | 3 - 4 | 296 | 98 | |
AK314916.1 | - | 4 - 5 | 465 | 98 | |
BC017569.1 | - | 4 - 5 | 490 | 98 | |
BC031073.1 | - | 4 - 5 | 552 | 98 | |
BX640960.1 | - | 3 - 4 | 367 | 98 | |
BX647487.1 | - | 4 - 5 | 483 | 98 | |
CR749294.1 | - | 4 - 5 | 471 | 98 |