The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007033.4 | NP_008964.3 | 5 - 6 | 558 | 73 |
XM_005244713.4 | XP_005244770.2 | 5 - 6 | 1379 | 73 | |
XM_006710306.2 | XP_006710369.1 | 6 - 7 | 640 | 73 | |
XM_011540542.1 | XP_011538844.1 | 6 - 7 | 578 | 73 | |
XM_011540543.1 | XP_011538845.1 | 5 - 6 | 426 | 73 | |
XM_017000131.1 | XP_016855620.1 | 5 - 6 | 1909 | 73 | |
XM_017000132.1 | XP_016855621.1 | 6 - 7 | 668 | 73 | |
XM_017000133.1 | XP_016855622.1 | 6 - 7 | 751 | 73 | |
XM_017000134.1 | XP_016855623.1 | 7 - 8 | 665 | 73 | |
XM_017000135.1 | XP_016855624.1 | 5 - 6 | 665 | 73 | |
GenBank mRNA | AF157324.1 | - | 5 - 6 | 497 | 73 |
AJ001421.1 | - | 4 - 5 | 367 | 73 | |
AK095750.1 | - | 4 - 5 | 2153 | 73 | |
AK289658.1 | - | 5 - 6 | 492 | 73 | |
BC004965.1 | - | 5 - 6 | 493 | 73 | |
BT007224.1 | - | 4 - 5 | 367 | 73 | |
CR456806.1 | - | 4 - 5 | 367 | 73 |