The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 17 - 18 | 2492 | 99 |
GenBank mRNA | AF021351.1 | - | 18 - 19 | 2359 | 99 |
AK026023.1 | - | 8 - 9 | 1066 | 99 | |
AK300329.1 | - | 17 - 18 | 2472 | 99 | |
AY091461.1 | - | 2 - 3 | 166 | 99 | |
BC041089.1 | - | 17 - 18 | 2453 | 99 |