The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003920.3 | NP_003911.2 | 2 - 3 | 263 | 87 |
XM_011538939.2 | XP_011537241.1 | 2 - 3 | 241 | 87 | |
GenBank mRNA | AB015597.1 | - | 1 - 2 | 95 | 87 |
AF098162.1 | - | 2 - 3 | 237 | 87 | |
AK296869.1 | - | 2 - 3 | 243 | 87 | |
BC031514.1 | - | 2 - 3 | 226 | 87 | |
BC039842.1 | - | 2 - 3 | 250 | 87 | |
BC050557.1 | - | 2 - 3 | 290 | 87 |