The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317172.1 | NP_001304101.1 | 3 - 4 | 467 | 164 |
NM_003836.6 | NP_003827.3 | 3 - 4 | 467 | 164 | |
GenBank mRNA | AK289384.1 | - | 3 - 4 | 437 | 164 |
AK297918.1 | - | 2 - 3 | 363 | 164 | |
AK313258.1 | - | 3 - 4 | 437 | 164 | |
BC007741.2 | - | 3 - 4 | 416 | 164 | |
BC013197.1 | - | 3 - 4 | 383 | 164 | |
BC014015.2 | - | 3 - 4 | 412 | 164 | |
DQ017081.1 | - | 3 - 4 | 263 | 164 | |
DQ126100.1 | - | 3 - 4 | 263 | 164 | |
DQ138965.1 | - | 3 - 4 | 263 | 164 | |
U15979.1 | - | 3 - 4 | 436 | 164 | |
U15981.1 | - | 3 - 4 | 436 | 164 | |
X17544.1 | - | 3 - 4 | 427 | 163 | |
Z12172.1 | - | 3 - 4 | 263 | 164 |