The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007207.5 | NP_009138.1 | 1 - 2 | 190 | 74 |
GenBank mRNA | AB026436.1 | - | 1 - 2 | 235 | 74 |
AF179212.1 | - | 1 - 2 | 74 | 74 | |
AK293287.1 | - | 1 - 2 | 104 | 74 | |
BC031405.1 | - | 1 - 2 | 89 | 74 | |
BC063826.1 | - | 1 - 2 | 84 | 74 | |
BG720911.1 | - | 1 - 2 | 195 | 74 | |
BP222019.1 | - | 1 - 2 | 188 | 74 | |
BQ223632.1 | - | 1 - 2 | 132 | 74 |