The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146213.1 | NP_001139685.2 | 11 - 12 | 1370 | 140 |
NM_001146214.1 | NP_001139686.1 | 12 - 13 | 1462 | 140 | |
NM_022771.4 | NP_073608.4 | 12 - 13 | 1421 | 140 | |
NR_027449.1 | 10 - 11 | 1159 | 140 | ||
XM_006719564.2 | XP_006719627.1 | 12 - 13 | 1403 | 140 | |
XM_006719565.3 | XP_006719628.1 | 12 - 13 | 1341 | 140 | |
XM_006719566.2 | XP_006719629.1 | 13 - 14 | 1552 | 140 | |
XM_006719567.2 | XP_006719630.1 | 11 - 12 | 1238 | 140 | |
XM_011538679.1 | XP_011536981.1 | 14 - 15 | 1478 | 140 | |
XM_011538680.1 | XP_011536982.1 | 12 - 13 | 1436 | 140 | |
XM_011538682.1 | XP_011536984.1 | 11 - 12 | 1240 | 140 | |
XM_017019824.1 | XP_016875313.1 | 13 - 14 | 1438 | 140 | |
XM_017019825.1 | XP_016875314.1 | 12 - 13 | 1457 | 140 | |
XM_017019826.1 | XP_016875315.1 | 12 - 13 | 1387 | 140 | |
XM_017019827.1 | XP_016875316.1 | 9 - 10 | 1061 | 140 | |
XM_017019828.1 | XP_016875317.1 | 10 - 11 | 1179 | 140 | |
GenBank mRNA | AB449901.1 | - | 11 - 12 | 1295 | 140 |
AK022147.1 | - | 11 - 12 | 1318 | 140 | |
AK225221.1 | - | 11 - 12 | 1299 | 140 | |
AK225747.1 | - | 11 - 12 | 1316 | 140 | |
AK292306.1 | - | 10 - 11 | 1090 | 140 | |
AK297626.1 | - | 12 - 13 | 1410 | 140 | |
AL833639.1 | - | 10 - 11 | 1098 | 140 | |
BC028352.2 | - | 12 - 13 | 1369 | 140 |