The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146213.1 | NP_001139685.2 | 15 - 16 | 1792 | 105 |
NM_001146214.1 | NP_001139686.1 | 16 - 17 | 1884 | 105 | |
NM_022771.4 | NP_073608.4 | 16 - 17 | 1843 | 105 | |
NR_027449.1 | 14 - 15 | 1581 | 105 | ||
XM_006719564.2 | XP_006719627.1 | 16 - 17 | 1825 | 105 | |
XM_006719565.3 | XP_006719628.1 | 16 - 17 | 1763 | 105 | |
XM_006719566.2 | XP_006719629.1 | 17 - 18 | 1974 | 105 | |
XM_006719567.2 | XP_006719630.1 | 15 - 16 | 1660 | 105 | |
XM_011538679.1 | XP_011536981.1 | 18 - 19 | 1900 | 105 | |
XM_011538680.1 | XP_011536982.1 | 16 - 17 | 1858 | 105 | |
XM_011538682.1 | XP_011536984.1 | 15 - 16 | 1662 | 105 | |
XM_017019824.1 | XP_016875313.1 | 17 - 18 | 1860 | 105 | |
XM_017019825.1 | XP_016875314.1 | 16 - 17 | 1879 | 105 | |
XM_017019826.1 | XP_016875315.1 | 16 - 17 | 1809 | 105 | |
XM_017019827.1 | XP_016875316.1 | 13 - 14 | 1483 | 105 | |
XM_017019828.1 | XP_016875317.1 | 14 - 15 | 1601 | 105 | |
GenBank mRNA | AB449901.1 | - | 15 - 16 | 1717 | 105 |
AK022147.1 | - | 15 - 16 | 1740 | 105 | |
AK225221.1 | - | 15 - 16 | 1721 | 105 | |
AK225747.1 | - | 15 - 16 | 1738 | 105 | |
AK292306.1 | - | 14 - 15 | 1512 | 105 | |
AK297626.1 | - | 16 - 17 | 1832 | 105 | |
AL157464.1 | - | 4 - 5 | 423 | 105 | |
AL833639.1 | - | 14 - 15 | 1520 | 105 | |
BC028352.2 | - | 16 - 17 | 1791 | 105 |